Assay Details
| Gene Symbol: Frem3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167898.1 | NP_001161370.1 | 5 - 6 | 5793 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frem3| Gene Symbol: Frem3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167898.1 | NP_001161370.1 | 5 - 6 | 5793 | 66 | |
Set Membership: |
|