Assay Details
Gene Symbol: Olfr411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146709.2 | NP_666920.2 | 1 | 746 | 102 | |
GenBank mRNA | BC138439.1 | - | 1 | 746 | 8861459 | 102 |
BC138440.1 | - | 1 | 746 | 8861460 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969924 | BC138440.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969924 | BC138439.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969924 | NM_146709.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163389 | BC138440.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163389 | BC138439.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116407 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163389 | BC138440.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163389 | BC138439.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969924 | BC138440.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969924 | BC138439.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969924 | NM_146709.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P191572 | BC138440.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P191572 | BC138439.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P191572 | NM_146709.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920022 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920021 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920029 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920030 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920026 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27919994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920025 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27920027 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116407 | - | - |
Whole Mouse Genome G4122A | A_51_P191572 | BC138440.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P191572 | BC138439.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P191572 | NM_146709.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116328 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116407 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09189571 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09189580 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09472242 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09189575 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09215359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27920046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27919941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09545937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09545941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919983 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09189580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09189571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09464915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09249849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27919994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09215355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27920004 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10388309 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10388308 | - | - |
Affymetrix Mouse Clariom S Array | PSR1100029403.mm.1 | NM_146709.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100029403.mm.1 | NM_146709.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100029403.mm.1 | TC1100003192.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 859186 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1570403 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001570403 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2654085 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2654085 | - | - |
mouse-6 v1.1 (Illumina) | scl13054.1.1_66-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1570403 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl13054.1.1_66-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570403 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2654085 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2654085 | - | - |
