Assay Details
| Gene Symbol: Olfr915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146785.2 | NP_666996.2 | 1 | 146 | 97 | |
| GenBank mRNA | BC111098.1 | - | 1 | 141 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017247 | NM_146785.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017247 | BC111098.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126392 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017247 | NM_146785.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017247 | BC111098.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242293 | NM_146785.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P242293 | BC111098.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435478 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126392 | - | - |
| Whole Mouse Genome G4122A | A_51_P242293 | NM_146785.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P242293 | BC111098.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125832 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104189 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126392 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07939970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07939967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07939972 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07939976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07939972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07580666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26435420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07580343 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592372 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592371 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900017172.mm.1 | NM_146785.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017172.mm.1 | NM_146785.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017172.mm.1 | TC0900002060.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 573292 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5130091 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777135 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777135 | - | - |
| mouse-6 v1.1 (Illumina) | scl36018.1_153-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130091 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36018.1_153-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2829495 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2777134 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2829495 | - | - |

Olfr915
Genomic Map