Assay Details
Gene Symbol: Olfr911-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146873.2 | NP_667084.2 | 1 | 746 | 100 | |
GenBank mRNA | BC127963.1 | - | 1 | 746 | 40134907 | 100 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148522 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435087 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435064 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435074 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435060 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435066 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435062 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435055 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435073 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435098 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435067 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435078 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435058 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435070 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435056 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26435054 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148522 | - | - |
Whole Mouse Genome G4122A | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P438990 | NM_146873.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117614 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148522 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07713946 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07713941 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07713933 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07713948 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07713940 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07713945 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07713936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26435094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07727009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07513917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26435088 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10584425 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10584424 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 678541 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 7000377 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007000377 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2671243 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2671243 | - | - |
mouse-6 v1.1 (Illumina) | scl37090.1.1_111-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7000377 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl37090.1.1_111-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7000377 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2671243 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2671243 | - | - |
