Assay Details
| Gene Symbol: Olfr1225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146891.2 | NP_667102.2 | 1 | 853 | 124 | |
| GenBank mRNA | BC145849.1 | - | 1 | 853 | 40131152 | 124 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr1225
Genomic Map