Assay Details
| Gene Symbol: Olfr325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207153.2 | NP_997036.2 | 1 | 954 | 98 | |
| GenBank mRNA | BC141015.1 | - | 1 | 978 | 9055627 | 98 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031001 | NM_207153.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2031001 | BC141015.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733392 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031001 | NM_207153.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2031001 | BC141015.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27831663 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103437 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09339303 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09192691 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09142470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27831651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09046652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09138357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09192691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09138359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27831658 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10386194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10386196 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386191 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386185 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386193 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376392 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386195 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100005573.mm.1 | NM_207153.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005573.mm.1 | NM_207153.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005573.mm.1 | TC1100000692.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100005572.mm.1 | NM_207153.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005572.mm.1 | NM_207153.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100005572.mm.1 | TC1100000692.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106840471 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7050594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106840471 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648301 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2648301 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230203 | - | - |
| mouse-6 v1.1 (Illumina) | GI_46391068-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0258261.1_328-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050594 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106840471 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258261.1_328-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050594 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106840471 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2648301 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2648301 | - | - |

Olfr325
Genomic Map