Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100000054.mm.1 |
NM_001159750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000054.mm.1 |
NM_001159750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000054.mm.1 |
NM_001159751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000054.mm.1 |
NM_001159751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000054.mm.1 |
NM_011541.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000054.mm.1 |
NM_011541.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000054.mm.1 |
TC0100003861.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000047.mm.1 |
NM_001159750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000047.mm.1 |
NM_001159750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000047.mm.1 |
NM_011541.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000047.mm.1 |
NM_011541.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000047.mm.1 |
TC0100003861.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000050.mm.1 |
NM_001159750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000050.mm.1 |
NM_001159750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000050.mm.1 |
NM_011541.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000050.mm.1 |
NM_011541.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000050.mm.1 |
TC0100003861.mm.1 |
MTA TC |