Assay Details
| Gene Symbol: BC080695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007579.3 | NP_001007580.1 | 356 | 101 | ||
| GenBank mRNA | BC080695.1 | - | 357 | 30458858 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BC080695
Genomic Map