| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10510071 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510078 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510069 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510114 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510073 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510070 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510075 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510115 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510083 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510074 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510116 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510082 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510077 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510079 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510080 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10510076 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10510081 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10510068 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10510072 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10510112 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400015098.mm.1 |
NM_001007579.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400015098.mm.1 |
NM_001007579.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400015098.mm.1 |
TC0400001831.mm.1 |
MTA TC |