| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100000970.mm.1 |
NM_001033904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000970.mm.1 |
NM_001033904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000970.mm.1 |
TC0100000140.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000959.mm.1 |
NM_183322.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000959.mm.1 |
NM_183322.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000959.mm.1 |
TC0100000137.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000973.mm.1 |
NM_001033904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000973.mm.1 |
NM_001033904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000973.mm.1 |
TC0100000140.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000960.mm.1 |
NM_183322.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000960.mm.1 |
NM_183322.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000960.mm.1 |
TC0100000137.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000972.mm.1 |
NM_001033904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000972.mm.1 |
NM_001033904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000972.mm.1 |
TC0100000140.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100000961.mm.1 |
NM_183322.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000961.mm.1 |
NM_183322.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100000961.mm.1 |
TC0100000137.mm.1 |
MTA TC |