Assay Details
| Gene Symbol: B3gnt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036740.2 | NP_001031817.1 | 1 - 2 | 98 | 69 | |
| GenBank mRNA | AK154220.1 | - | 85 | 69 | ||
| AK154901.1 | - | 1 - 2 | 54 | 69 | ||
| BC025206.1 | - | 1 - 2 | 84 | 5037295 | 69 | |
| BY320287.1 | - | 1 - 2 | 82 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154220.1, AK154901.1, BC025206.1, NM_001036740.2 transcripts.
Set Membership: |
|

B3gnt8
Genomic Map