| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300013208.mm.1 |
NM_001076679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013208.mm.1 |
NM_001076679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013208.mm.1 |
TC0300001673.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300013210.mm.1 |
NM_001076679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013210.mm.1 |
NM_001076679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013210.mm.1 |
TC0300001673.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300013209.mm.1 |
NM_001076679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013209.mm.1 |
NM_001076679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013209.mm.1 |
TC0300001673.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300013211.mm.1 |
NM_001076679.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013211.mm.1 |
NM_001076679.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300013211.mm.1 |
TC0300001673.mm.1 |
MTA TC |