Assay Details
| Gene Symbol: Prrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC060135.1 | - | 3 - 4 | 229 | 80 | |
| BC137616.1 | - | 3 - 4 | 358 | 8860635 | 80 | |
| BC137619.1 | - | 3 - 4 | 358 | 8860638 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P293120 | NM_001081135.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P293120 | AK079502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P293120 | BC060135.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P293120 | NM_001081135.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P293120 | AK079502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P293120 | BC060135.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500870 | AK076442.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500870 | AK087423.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500870 | AK163823.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500870 | BC031924.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500870 | L77867.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500870 | NM_007680.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19661 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127780 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172115 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143932 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P293120 | NM_001081135.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P293120 | AK079502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P293120 | BC060135.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P293120 | NM_001081135.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P293120 | AK079502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P293120 | BC060135.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172115 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09326025 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143932 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127780 | - | - |
| Whole Mouse Genome G4122A | A_52_P293120 | NM_001081135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293120 | AK079502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293120 | BC060135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293120 | NM_001081135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293120 | AK079502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293120 | BC060135.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127780 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143932 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14774959 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14774972 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14774979 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14774978 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14774967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32456144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14621113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14791941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14531485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14774981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14627015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14774953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15200539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14635727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14748641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14635733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14751109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14748653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14531493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14531478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15126892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14661035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14774967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32456184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14656124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15217680 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10600045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600046 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439549_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600044 | - | - |
| Mouse Genome 430 2.0 Array | 1439549_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439549_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208748_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208748_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00004140.mm.1 | NM_001081135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004140.mm.1 | TC0X00000660.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004146.mm.1 | NM_001081135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004146.mm.1 | TC0X00000660.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004144.mm.1 | NM_001081135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004144.mm.1 | TC0X00000660.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004141.mm.1 | NM_001081135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004141.mm.1 | TC0X00000660.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004142.mm.1 | NM_001081135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004142.mm.1 | TC0X00000660.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004138.mm.1 | TC0X00000660.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004145.mm.1 | NM_001081135.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004145.mm.1 | TC0X00000660.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439549_at | - | - |
| Mouse Expression Set 430 | 1439549_PM_at | - | - |
| Murine Genome U74v2 Set | 133337_PM_at | - | - |
| Murine Genome U74v2 Set | 133337_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439549_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439549_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241345 | - | - |

Prrg3
Genomic Map