Assay Details
Gene Symbol: Ttc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081228.1 | NP_001074697.1 | 1 | 553 | 102 | |
GenBank mRNA | BC157934.1 | - | 1 | 574 | 9088113 | 102 |
BC158130.1 | - | 1 | 670 | 9007398 | 102 | |
BC172090.1 | - | 1 | 574 | 9054376 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2080378 | BC157934.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2080378 | BC158130.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2080378 | BC172090.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2080378 | NM_001081228.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2080378 | BC157934.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2080378 | BC158130.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2080378 | BC172090.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2080378 | NM_001081228.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02126155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01639478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01693179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347180 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01899649 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01574157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01574159 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01714088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01742406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02126157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01714084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21347231 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10483823 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437098_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455921_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10483822 | - | - |
Mouse Genome 430 2.0 Array | 1437098_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1455921_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455921_at | - | - |
Mouse Genome 430 2.0 Array | 1437098_x_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200032431.mm.1 | TC0200003799.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200032432.mm.1 | TC0200003799.mm.1 | MTA TC |
Mouse Expression Set 430 | 1437098_PM_x_at | - | - |
Mouse Expression Set 430 | 1437098_x_at | - | - |
Mouse Expression Set 430 | 1455921_PM_at | - | - |
Mouse Expression Set 430 | 1455921_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455921_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437098_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455921_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437098_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_3014466 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3014466 | - | - |
