| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00000747.mm.1 |
NM_001081564.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000747.mm.1 |
NM_001081564.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000747.mm.1 |
TC0X00000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000748.mm.1 |
NM_001081564.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000748.mm.1 |
NM_001081564.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000748.mm.1 |
TC0X00000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000749.mm.1 |
NM_001081564.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000749.mm.1 |
NM_001081564.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000749.mm.1 |
TC0X00000094.mm.1 |
MTA TC |