Assay Details
| Gene Symbol: Vmn2r31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105062.1 | NP_001098532.1 | 3 - 4 | 1303 | 101 | |
| Gene Symbol: Vmn2r44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105074.1 | NP_001098544.1 | 3 - 4 | 1303 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vmn2r31
Genomic Map