Assay Details
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110149.1 | NP_001103619.1 | 1 - 2 | 54 | 109 | |
GenBank mRNA | AK040121.1 | - | 1 - 2 | 55 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK040121.1, NM_001110149.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
