Assay Details
| Gene Symbol: Rpl18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029751.4 | NP_084027.1 | 3 - 4 | 400 | 97 | |
| GenBank mRNA | AK010983.1 | - | 3 - 4 | 371 | 97 | |
| AK146543.1 | - | 3 - 4 | 370 | 97 | ||
| AK150820.1 | - | 3 - 4 | 369 | 97 | ||
| AK152611.1 | - | 3 - 4 | 98 | |||
| AK161942.1 | - | 3 - 4 | 370 | 97 | ||
| AK162047.1 | - | 3 - 4 | 371 | 97 | ||
| AK168258.1 | - | 3 - 4 | 370 | 97 | ||
| BC024609.1 | - | 3 - 4 | 344 | 4221597 | 97 | |
| BC037146.1 | - | 3 - 4 | 412 | 5011713 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl18a
Genomic Map