Assay Details
| Gene Symbol: 9930111J21Rik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114679.1 | NP_001108151.1 | 82 | 107 | ||
| Gene Symbol: 9930111J21Rik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC066104.1 | - | 82 | 107 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001114679.1 transcripts.
This assay is in the 5'UTR region of the NM_001114679.1 transcripts.
Set Membership: |
|

9930111J21Rik1
Genomic Map