| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10385530 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10385517 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10385532 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10385515 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10385516 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10385531 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10385526 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10385513 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1100024165.mm.1 |
NM_001024230.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024165.mm.1 |
NM_001024230.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024165.mm.1 |
TC1100002642.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040883.mm.1 |
NM_001114679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040883.mm.1 |
NM_001114679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040883.mm.1 |
TC1100004292.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1425728_PM_at |
- |
- |
| Mouse Expression Set 430 |
1425728_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1425728_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1425728_at |
- |
- |