Assay Details
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020567.2 | NP_065592.1 | 3 - 4 | 258 | 90 | |
GenBank mRNA | AF068780.1 | - | 3 - 4 | 405 | 90 | |
AK131959.1 | - | 3 - 4 | 232 | 90 | ||
BC020061.1 | - | 3 - 4 | 410 | 3988065 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
