Assay Details
| Gene Symbol: Rsc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023544.5 | NP_076033.4 | 1 | 52 | 110 | |
| GenBank mRNA | AI854132.1 | - | 1 | 99 | 110 | |
| BC150752.1 | - | 1 | 52 | 9087663 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC150752.1,NM_023544.5 transcripts.
This assay is in the 5'UTR region of the BC150752.1,NM_023544.5 transcripts.
Set Membership: |
|

Rsc1a1
Genomic Map