Assay Details
| Gene Symbol: Olfr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146465.2 | NP_666676.1 | 1 | 46 | 75 | |
| GenBank mRNA | BC132008.1 | - | 1 | 46 | 40130285 | 75 |
| BC132012.1 | - | 1 | 46 | 40130289 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC132008.1,BC132012.1,NM_146465.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr108
Genomic Map