Assay Details
| Gene Symbol: Pln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141927.1 | NP_001135399.1 | 2 - 3 | 233 | 106 | |
| GenBank mRNA | CA459471.1 | - | 2 - 3 | 226 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001141927.1 transcripts.
Set Membership: |
|

Pln
Genomic Map