Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10414247 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10419164 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414219 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10419163 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414227 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414237 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414233 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414231 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414240 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414220 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414221 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10419166 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414249 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10414230 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10419162 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10414245 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1400014254.mm.1 |
NM_001162954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014254.mm.1 |
NM_001162954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400014254.mm.1 |
TC1400001888.mm.1 |
MTA TC |