Assay Details
Gene Symbol: Serpinb6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164118.1 | NP_001157590.1 | 2 - 3 | 385 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001164118.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
