Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016695.mm.1 |
NM_199015.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016695.mm.1 |
NM_199015.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016695.mm.1 |
TC1400002869.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016692.mm.1 |
NM_199015.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016692.mm.1 |
NM_199015.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016692.mm.1 |
TC1400002869.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016693.mm.1 |
NM_199015.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016693.mm.1 |
NM_199015.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016693.mm.1 |
TC1400002869.mm.1 |
MTA TC |