Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200014827.mm.1 |
NM_001166062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014827.mm.1 |
NM_001166062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014827.mm.1 |
TC1200002128.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200014826.mm.1 |
NM_001166062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014826.mm.1 |
NM_001166062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014826.mm.1 |
TC1200002128.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200014828.mm.1 |
NM_001166062.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014828.mm.1 |
NM_001166062.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014828.mm.1 |
TC1200002128.mm.1 |
MTA TC |