Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100037811.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037811.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037811.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037811.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037811.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037799.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037799.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037799.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037799.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037799.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037797.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037797.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037797.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037797.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037797.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037803.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037808.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037808.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037808.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037808.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037808.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037816.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037816.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037816.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037816.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037816.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037798.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037798.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037798.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037798.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037798.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037805.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037805.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037805.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037805.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037805.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037796.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037796.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037796.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037796.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037796.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037802.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037802.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037802.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037802.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037802.mm.1 |
TC1100004032.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100037804.mm.1 |
NM_001166503.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037804.mm.1 |
NM_001166503.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037804.mm.1 |
NM_027216.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037804.mm.1 |
NM_027216.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100037804.mm.1 |
TC1100004032.mm.1 |
MTA TC |