Assay Details
| Gene Symbol: Serpina3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167705.1 | NP_001161177.1 | 3 - 4 | 1134 | 93 | |
| NM_028740.2 | NP_083016.1 | 2 - 3 | 474 | 93 | ||
| GenBank mRNA | AK016702.1 | - | 2 - 3 | 477 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serpina3a
Genomic Map