Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_001172472.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_001172472.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_001172473.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_001172473.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_001172475.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_001172475.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_011451.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_011451.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_025367.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
NM_025367.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018383.mm.1 |
TC1100001935.mm.1 |
MTA TC |