Assay Details
| Gene Symbol: Ear2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007895.2 | NP_031921.1 | 1 - 2 | 69 | 79 | |
| GenBank mRNA | AF306665.1 | - | 1 - 2 | 69 | 79 | |
| BC065391.1 | - | 1 - 2 | 40 | 3464790 | 79 | |
| Gene Symbol: Ear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007894.2 | NP_031920.1 | 66 | 79 | ||
| GenBank mRNA | AY316149.1 | - | 66 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AF306665.1,AY316149.1,BC065391.1,NM_007894.2,NM_007895.2 transcripts.
This assay is in the 5'UTR region of the AF306665.1,AY316149.1,BC065391.1,NM_007894.2,NM_007895.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952618 | AF306665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952618 | BC065391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952618 | BC094626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952618 | NM_007895.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952618 | NM_007894.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | BC117060.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | U72032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | NM_007894.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | AY316149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | BC117060.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | U72032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | NM_007894.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412585 | AY316149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952618 | AF306665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952618 | BC065391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952618 | BC094626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952618 | NM_007895.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952618 | NM_007894.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | BC117060.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | U72032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | NM_007894.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | AY316149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | BC117060.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | U72032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | NM_007894.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412585 | AY316149.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29566550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29566557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29566563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29566571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29566567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29566553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29566537 | - | - |
| Whole Mouse Genome G4122A | A_52_P412585 | BC117060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412585 | U72032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412585 | NM_007894.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412585 | AY316149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412585 | BC117060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412585 | U72032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412585 | NM_007894.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412585 | AY316149.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131103 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129393 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29566573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29566550 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10419153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414264 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10419157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414263 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598177 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421802_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449846_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419156 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598175 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414262 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419151 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419154 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419154 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10419156 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598175 | - | - |
| Mouse Genome 430 2.0 Array | 1449846_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421802_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449846_at | - | - |
| Mouse Genome 430 2.0 Array | 1421802_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13586_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13587_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13587_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13586_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400003787.mm.1 | NM_007895.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003787.mm.1 | NM_007895.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003787.mm.1 | TC1400000459.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400003781.mm.1 | NM_007895.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003781.mm.1 | NM_007895.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400003781.mm.1 | TC1400000459.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400014173.mm.1 | NM_007894.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014173.mm.1 | NM_007894.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400014173.mm.1 | TC1400001868.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422411_PM_s_at | - | - |
| Mouse Expression Set 430 | 1421802_PM_at | - | - |
| Mouse Expression Set 430 | 1422411_s_at | - | - |
| Mouse Expression Set 430 | 1422411_PM_s_at | - | - |
| Mouse Expression Set 430 | 1421802_at | - | - |
| Mouse Expression Set 430 | 1449846_PM_at | - | - |
| Mouse Expression Set 430 | 1449846_at | - | - |
| Mouse Expression Set 430 | 1422411_s_at | - | - |
| Murine Genome U74v2 Set | 103240_f_at | - | - |
| Murine Genome U74v2 Set | 103240_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100311_PM_f_at | - | - |
| Murine Genome U74v2 Set | 100311_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422411_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421802_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449846_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422411_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421802_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422411_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422411_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449846_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421802_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421802_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449846_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449846_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 685503 | - | - |
| ABI Mouse Genome Survey Microarray | 925991 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5670239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106620332 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2360471 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1340239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005670239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360471 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2731735 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232396 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251292 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2731735 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232396 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2524992 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251292 | - | - |
| mouse-6 v1.1 (Illumina) | GI_34328111-S | - | - |
| mouse-6 v1.1 (Illumina) | scl013586.1_302-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0013586.1_285-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0013587.1_32-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340239 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106620332 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360471 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5670239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013587.1_32-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013586.1_285-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013586.1_302-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360471 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5670239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106620332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1232396 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232396 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2524992 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2731735 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2925653 | - | - |

Ear2
Genomic Map