Assay Details
Gene Symbol: Gpx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008160.6 | NP_032186.2 | 1 - 2 | 482 | 78 | |
GenBank mRNA | AK002245.1 | - | 1 - 2 | 287 | 78 | |
AK010999.1 | - | 1 - 2 | 287 | 78 | ||
AK011019.1 | - | 1 - 2 | 287 | 78 | ||
AK028171.1 | - | 1 - 2 | 287 | 78 | ||
AK150548.1 | - | 1 - 2 | 288 | 78 | ||
AK154833.1 | - | 1 - 2 | 287 | 78 | ||
AK160388.1 | - | 1 - 2 | 288 | 78 | ||
BC086649.1 | - | 1 - 2 | 270 | 6819448 | 78 | |
CK628430.1 | - | 1 - 2 | 482 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
temperature homeostasis
endothelial cell development
negative regulation of inflammatory response to antigenic stimulus
lipid metabolic process
triglyceride metabolic process
glutathione metabolic process
apoptotic process
response to oxidative stress
response to lipid hydroperoxide
sensory perception of sound
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
response to xenobiotic stimulus
response to symbiotic bacterium
response to wounding
response to toxic substance
UV protection
response to selenium ion
response to gamma radiation
myotube differentiation
protein oxidation
response to hydroperoxide
regulation of mammary gland epithelial cell proliferation
regulation of gene expression, epigenetic
vasodilation
response to hydrogen peroxide
hydrogen peroxide catabolic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
skeletal muscle tissue regeneration
regulation of neuron apoptotic process
blood vessel endothelial cell migration
fat cell differentiation
cell redox homeostasis
skeletal muscle fiber development
myoblast proliferation
interaction with symbiont
positive regulation of protein kinase B signaling
oxidation-reduction process
heart contraction
angiogenesis involved in wound healing
regulation of proteasomal protein catabolic process
negative regulation of release of cytochrome c from mitochondria
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
peroxidase activity
glutathione peroxidase activity
protein binding
selenium binding
oxidoreductase activity
SH3 domain binding
glutathione binding
phospholipid-hydroperoxide glutathione peroxidase activity
response to reactive oxygen species
temperature homeostasis
endothelial cell development
negative regulation of inflammatory response to antigenic stimulus
lipid metabolic process
triglyceride metabolic process
glutathione metabolic process
apoptotic process
response to oxidative stress
response to lipid hydroperoxide
sensory perception of sound
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
response to xenobiotic stimulus
response to symbiotic bacterium
response to wounding
response to toxic substance
UV protection
response to selenium ion
response to gamma radiation
myotube differentiation
protein oxidation
response to hydroperoxide
regulation of mammary gland epithelial cell proliferation
regulation of gene expression, epigenetic
vasodilation
response to hydrogen peroxide
hydrogen peroxide catabolic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
skeletal muscle tissue regeneration
regulation of neuron apoptotic process
blood vessel endothelial cell migration
fat cell differentiation
cell redox homeostasis
skeletal muscle fiber development
myoblast proliferation
interaction with symbiont
positive regulation of protein kinase B signaling
oxidation-reduction process
heart contraction
angiogenesis involved in wound healing
regulation of proteasomal protein catabolic process
negative regulation of release of cytochrome c from mitochondria
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
peroxidase activity
glutathione peroxidase activity
protein binding
selenium binding
oxidoreductase activity
SH3 domain binding
glutathione binding
phospholipid-hydroperoxide glutathione peroxidase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814365 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814368 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814364 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814363 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814362 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814367 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814366 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK028171.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK150548.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK154833.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK160388.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | BC086649.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK028171.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK150548.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK154833.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | AK160388.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P684378 | BC086649.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151026 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK028171.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK150548.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK154833.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK160388.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | BC086649.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK028171.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK150548.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK154833.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | AK160388.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P684378 | BC086649.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK028171.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK150548.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK154833.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK160388.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | BC086649.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK028171.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK150548.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK154833.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | AK160388.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P684378 | BC086649.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814359 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814369 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814358 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814367 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814370 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814353 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814362 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814365 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814361 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814368 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26814356 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151026 | - | - |
Whole Mouse Genome G4122A | A_52_P684378 | AK028171.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | AK150548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | AK154833.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | AK160388.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | BC086649.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | AK028171.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | AK150548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | AK154833.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | AK160388.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P684378 | BC086649.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136272 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151026 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08029076 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08029062 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08029067 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08029086 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08102791 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08102790 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814365 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814367 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814366 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814363 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814364 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814362 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26814368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26814411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08029066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08102791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26814369 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10588900 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588902 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10588901 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1460671_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10588899 | - | - |
Mouse Genome 430 2.0 Array | 1460671_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1460671_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14775_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14775_PM_at | - | - |
Mouse Expression Set 430 | 1460671_at | - | - |
Mouse Expression Set 430 | 1460671_PM_at | - | - |
Murine Genome U74v2 Set | 94132_PM_at | - | - |
Murine Genome U74v2 Set | 94132_at | - | - |
Mouse Genome 430A 2.0 Array | 1460671_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1460671_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460671_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460671_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4150093 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004150093 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241367 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1241367 | - | - |
mouse-6 v1.1 (Illumina) | scl36472.2.242_20-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4150093 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl36472.2.242_20-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150093 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1241367 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241367 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36472.2.242_20-S | - | - |
