Assay Details
| Gene Symbol: Hsd3b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008295.2 | NP_032321.2 | 1 - 2 | 57 | 95 | |
| GenBank mRNA | BC012715.1 | - | 1 - 2 | 61 | 4193717 | 95 |
| BY097811.1 | - | 1 - 2 | 140 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC012715.1, NM_008295.2 transcripts.
Set Membership: |
|

Hsd3b5
Genomic Map