Assay Details
| Gene Symbol: Ins1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008386.3 | NP_032412.3 | 1 - 2 | 171 | 92 | |
| GenBank mRNA | BP773571.1 | - | 1 - 2 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_008386.3 transcripts.
Set Membership: |
|

Ins1
Genomic Map