Assay Details
| Gene Symbol: Mageb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008545.2 | NP_032571.2 | 1 | 752 | 96 | |
| GenBank mRNA | AK019658.1 | - | 1 | 752 | 96 | |
| BC141002.1 | - | 1 | 650 | 9055614 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK019658.1,BC141002.1,NM_008545.2 transcripts.
This assay is in the 5'UTR region of the AK019658.1,BC141002.1,NM_008545.2 transcripts.
Set Membership: |
|

Mageb3
Genomic Map