Assay Details
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008568.2 | NP_032594.1 | 12 - 13 | 1784 | 73 | |
GenBank mRNA | AK076059.1 | - | 8 - 9 | 1234 | 73 | |
AK077766.1 | - | 9 - 10 | 1343 | 73 | ||
AK132748.1 | - | 2348 | 73 | |||
AK133091.1 | - | 12 - 13 | 1780 | 73 | ||
AK144980.1 | - | 12 - 13 | 1796 | 73 | ||
AK149913.1 | - | 12 - 13 | 1778 | 73 | ||
AK150059.1 | - | 12 - 13 | 1781 | 73 | ||
AK150066.1 | - | 12 - 13 | 1778 | 73 | ||
AK154052.1 | - | 12 - 13 | 1781 | 73 | ||
BC023232.1 | - | 12 - 13 | 1760 | 73 | ||
BC065164.1 | - | 12 - 13 | 1768 | 6829737 | 73 | |
BC066024.1 | - | 12 - 13 | 1723 | 6829337 | 73 | |
D26091.1 | - | 12 - 13 | 1784 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
