Assay Details
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008568.2 | NP_032594.1 | 8 - 9 | 1089 | 97 | |
GenBank mRNA | AK076059.1 | - | 4 - 5 | 539 | 97 | |
AK077766.1 | - | 5 - 6 | 651 | 97 | ||
AK132748.1 | - | 1650 | 97 | |||
AK133091.1 | - | 8 - 9 | 1085 | 97 | ||
AK144980.1 | - | 8 - 9 | 1101 | 97 | ||
AK149913.1 | - | 8 - 9 | 1083 | 97 | ||
AK150059.1 | - | 8 - 9 | 1086 | 97 | ||
AK150066.1 | - | 8 - 9 | 1083 | 97 | ||
AK154052.1 | - | 8 - 9 | 1086 | 97 | ||
BC023232.1 | - | 8 - 9 | 1065 | 97 | ||
BC065164.1 | - | 8 - 9 | 1073 | 6829737 | 97 | |
BC066024.1 | - | 8 - 9 | 1028 | 6829337 | 97 | |
D26091.1 | - | 8 - 9 | 1089 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
