Assay Details
Gene Symbol: Pip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008843.3 | NP_032869.2 | 2 - 3 | 224 | 112 | |
GenBank mRNA | AB017907.1 | - | 2 - 3 | 202 | 112 | |
AK138795.1 | - | 2 - 3 | 225 | 112 | ||
BC028889.1 | - | 2 - 3 | 222 | 4165968 | 112 | |
S73282.1 | - | 2 - 3 | 220 | 112 | ||
X02510.1 | - | 2 - 3 | 219 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
