Assay Details
| Gene Symbol: Pspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008954.2 | NP_032980.2 | 1 - 2 | 159 | 90 | |
| GenBank mRNA | AF040960.1 | - | 1 - 2 | 156 | 90 | |
| BC130229.1 | - | 1 - 2 | 160 | 40141900 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pspn
Genomic Map