Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700000554.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000554.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000554.mm.1 |
TC1700000073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000555.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000555.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000555.mm.1 |
TC1700000073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001163.mm.1 |
NM_009341.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001163.mm.1 |
NM_009341.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001163.mm.1 |
TC1700002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001158.mm.1 |
NM_009341.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001158.mm.1 |
NM_009341.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001158.mm.1 |
TC1700002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001157.mm.1 |
NM_009341.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001157.mm.1 |
NM_009341.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001157.mm.1 |
TC1700002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001165.mm.1 |
NM_009341.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001165.mm.1 |
NM_009341.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001165.mm.1 |
TC1700002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000558.mm.1 |
NM_011553.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000558.mm.1 |
NM_011553.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000558.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000558.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000558.mm.1 |
TC1700000073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000561.mm.1 |
NM_011553.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000561.mm.1 |
NM_011553.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000561.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000561.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000561.mm.1 |
TC1700000073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000562.mm.1 |
NM_011553.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000562.mm.1 |
NM_011553.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000562.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000562.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000562.mm.1 |
TC1700000073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001180.mm.1 |
NM_001167578.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001180.mm.1 |
NM_001167578.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001180.mm.1 |
TC1700002767.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001182.mm.1 |
NM_001167578.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001182.mm.1 |
NM_001167578.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001182.mm.1 |
TC1700002767.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001162.mm.1 |
NM_009341.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001162.mm.1 |
NM_009341.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001162.mm.1 |
TC1700002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001181.mm.1 |
NM_001167578.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001181.mm.1 |
NM_001167578.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001181.mm.1 |
TC1700002767.mm.1 |
MTA TC |