Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700001170.mm.1 |
NM_009341.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001170.mm.1 |
NM_009341.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001170.mm.1 |
TC1700002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700001171.mm.1 |
NM_009341.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001171.mm.1 |
NM_009341.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700001171.mm.1 |
TC1700002766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000570.mm.1 |
NM_011553.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000570.mm.1 |
NM_011553.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000570.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000570.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000570.mm.1 |
TC1700000073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000571.mm.1 |
NM_011553.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000571.mm.1 |
NM_011553.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000571.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000571.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000571.mm.1 |
TC1700000073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000569.mm.1 |
NM_011553.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000569.mm.1 |
NM_011553.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000569.mm.1 |
NR_046291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000569.mm.1 |
NR_046291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000569.mm.1 |
TC1700000073.mm.1 |
MTA TC |