Assay Details
| Gene Symbol: Bcl7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009746.2 | NP_033876.1 | 3 - 4 | 523 | 85 | |
| GenBank mRNA | AK131935.1 | - | 3 - 4 | 412 | 85 | |
| BC005673.1 | - | 3 - 4 | 523 | 3153237 | 85 | |
| BC058530.1 | - | 3 - 4 | 394 | 6818556 | 85 | |
| Y11905.1 | - | 4 - 5 | 341 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcl7c
Genomic Map