Assay Details
| Gene Symbol: Gtf2h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF054823.1 | - | 7 - 8 | 802 | 93 | |
| AK079456.1 | - | 7 - 8 | 831 | 93 | ||
| AK088296.1 | - | 7 - 8 | 828 | 93 | ||
| AK144680.1 | - | 7 - 8 | 833 | 93 | ||
| AK146112.1 | - | 7 - 8 | 832 | 93 | ||
| AK149320.1 | - | 7 - 8 | 833 | 93 | ||
| BC012638.1 | - | 6 - 7 | 691 | 4191809 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gtf2h4
Genomic Map