Assay Details
Gene Symbol: Prl2c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011118.2 | NP_035248.2 | 1 - 2 | 99 | 87 | |
GenBank mRNA | AK145995.1 | - | 1 - 2 | 101 | 87 | |
AK164068.1 | - | 1 - 2 | 100 | 87 | ||
BC132078.1 | - | 1 - 2 | 81 | 40130355 | 87 | |
BC132080.1 | - | 1 - 2 | 81 | 40130357 | 87 | |
K03235.1 | - | 1 - 2 | 98 | 87 |
Gene Symbol: Prl2c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011954.2 | NP_036084.2 | 89 | 87 | ||
GenBank mRNA | BC058816.1 | - | 89 | 30053850 | 87 | |
BC064772.1 | - | 80 | 30434146 | 87 | ||
BC100299.1 | - | 71 | 30713114 | 87 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK145995.1,AK164068.1,BC058816.1,BC064772.1,BC100299.1,BC132078.1,BC132080.1,K03235.1,NM_011118.2,NM_011954.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
