Assay Details
Gene Symbol: Prl2c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011118.2 | NP_035248.2 | 2 - 3 | 269 | 88 | |
GenBank mRNA | AK145995.1 | - | 2 - 3 | 271 | 88 | |
AK164068.1 | - | 2 - 3 | 270 | 88 | ||
BC132078.1 | - | 2 - 3 | 251 | 40130355 | 88 | |
BC132080.1 | - | 2 - 3 | 251 | 40130357 | 88 | |
K03235.1 | - | 2 - 3 | 268 | 88 |
Gene Symbol: Prl2c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011954.2 | NP_036084.2 | 259 | 88 | ||
GenBank mRNA | BC058816.1 | - | 259 | 30053850 | 88 | |
BC064772.1 | - | 250 | 30434146 | 88 | ||
BC100299.1 | - | 241 | 30713114 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
