Assay Details
Gene Symbol: Saa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011316.3 | NP_035446.1 | 1 - 2 | 112 | 102 | |
GenBank mRNA | BC019212.1 | - | 1 - 2 | 148 | 5053373 | 102 |
U02554.1 | - | 1 - 2 | 143 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC019212.1, NM_011316.3, U02554.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
