Assay Details
Gene Symbol: C4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011413.2 | NP_035543.2 | 1207 | 109 | ||
GenBank mRNA | AK129017.1 | - | 1217 | 109 | ||
BC141050.1 | - | 1178 | 9055662 | 109 | ||
M21576.1 | - | 1209 | 109 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
