Assay Details
| Gene Symbol: Serpinb9g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011455.3 | NP_035585.2 | 4 - 5 | 496 | 109 | |
| GenBank mRNA | AF425083.1 | - | 4 - 5 | 491 | 109 | |
| AK005394.1 | - | 4 - 5 | 496 | 109 | ||
| BC064758.1 | - | 4 - 5 | 481 | 30018322 | 109 | |
| BC064759.1 | - | 4 - 5 | 471 | 30432705 | 109 | |
| Gene Symbol: Serpinb9f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183197.1 | NP_899020.1 | 4 - 5 | 485 | 109 | |
| GenBank mRNA | BC052216.1 | - | 4 - 5 | 485 | 30020424 | 109 |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb9g
Genomic Map