Assay Details
| Gene Symbol: Eif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010579.2 | NP_034709.1 | 3 - 4 | 855 | 104 | |
| GenBank mRNA | AF047046.1 | - | 3 - 4 | 594 | 104 | |
| AF108462.2 | - | 3 - 4 | 640 | 104 | ||
| AK003447.1 | - | 3 - 4 | 611 | 104 | ||
| AK007594.1 | - | 3 - 4 | 622 | 104 | ||
| AK012218.1 | - | 3 - 4 | 621 | 104 | ||
| AK088707.1 | - | 3 - 4 | 618 | 104 | ||
| AK134364.1 | - | 3 - 4 | 622 | 104 | ||
| AK150400.1 | - | 3 - 4 | 629 | 104 | ||
| AK150720.1 | - | 3 - 4 | 628 | 104 | ||
| AK150931.1 | - | 3 - 4 | 630 | 104 | ||
| AK151089.1 | - | 3 - 4 | 630 | 104 | ||
| AK151102.1 | - | 3 - 4 | 613 | 104 | ||
| AK151559.1 | - | 3 - 4 | 609 | 104 | ||
| AK151644.1 | - | 3 - 4 | 629 | 104 | ||
| AK152093.1 | - | 3 - 4 | 614 | 104 | ||
| AK152215.1 | - | 3 - 4 | 614 | 104 | ||
| AK152416.1 | - | 3 - 4 | 636 | 104 | ||
| AK159157.1 | - | 3 - 4 | 614 | 104 | ||
| AK165946.1 | - | 3 - 4 | 413 | 104 | ||
| AK167422.1 | - | 3 - 4 | 609 | 104 | ||
| AK168257.1 | - | 3 - 4 | 630 | 104 | ||
| BC015274.1 | - | 3 - 4 | 855 | 4219358 | 104 | |
| BC024442.1 | - | 3 - 4 | 583 | 4973875 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif6
Genomic Map